Reagents Ammonium Chloride, Purified, Poly Pail, 10 kg

Representative image only.

Reagents Ammonium Chloride, Purified, Poly Pail, 10 kg

Reagents | Mfr: C2132000-10KG| Item: 6228387

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White solid
  • Density: 1.53
  • Melting point: 337.8 °C (640.04 °F) with decomposition
  • Solubility information: 395 g/L at 25 °C (77 °F)
  • pH range: 4.5 to 5.5
  • 10 kg poly pail

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNH4Cl
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)10000
  • Formula Weight53.49
  • Vapor Pressure (torr)0.9976
  • PackagingPoly pail
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number12125-02-9
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)520
  • Boiling Point F (° F)968
  • DescriptionAmmonium Chloride, Purified, Poly Pail, 10 kg
Documents & Certificates
Reviews
Q&A