Reagents Ammonium Carbonate, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Ammonium Carbonate, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2131500-2.5KG| Item: 6228386

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • White solid
  • Odor: strong, ammonia-like
  • Density: 1.5
  • Boiling point: 60 °C (140 °F), decomposes
  • Solubility information: miscible
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear Formula(NH4)2CO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)2500
  • Formula Weight96.09
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number506-87-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)58
  • Melting Point F (° F)136.4
  • DescriptionAmmonium Carbonate, Purified, Natural Poly Container, 2.5 kg
Documents & Certificates
Reviews
Q&A