Reagents Acetone, Purified, Drum, 200 L

Representative image only.

Reagents Acetone, Purified, Drum, 200 L

Reagents | Mfr: C2102500-200E| Item: 6228337

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.8
  • Solubility information: miscible
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • Synonyms2-Propanone, Dimethyl ketone
  • FormLiquid
  • Volume (mL)200000
  • Formula Weight58.08
  • Vapor Pressure (torr)174.764
  • PackagingDrum
  • DOT InformationACETONE, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1090
  • CAS Number67-64-1
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, Purified, Drum, 200 L
Documents & Certificates
Reviews
Q&A