Reagents Acetone, Meets ASTM D 329, Poly Bottle, 4 L

Representative image only.

Reagents Acetone, Meets ASTM D 329, Poly Bottle, 4 L

Reagents | Mfr: C1102550-4A| Item: 6228113

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • ASTMD 329 certified
  • Colorless liquid
  • Flash point: -20 °C (-4 °F)
  • Density: 0.8
  • 4 L poly bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeASTM
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Propanone, Dimethyl ketone
  • Volume (mL)4000
  • Formula Weight58.08
  • Vapor Pressure (torr)176.7644
  • PackagingPoly bottle
  • DOT InformationACETONE, 3
  • Storage SymbolAmbient
  • CAS Number67-64-1
  • UN NumberUN1090
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, Meets ASTM D 329, Poly Bottle, 4 L
Documents & Certificates
Reviews
Q&A