Reagents Acetone, ACS Reagent, Drum, 200 L

Representative image only.

Reagents Acetone, ACS Reagent, Drum, 200 L

Reagents | Mfr: C1102500-200E| Item: 6228111

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price

Buy It Together

Current Item

Reagents Acetone, ACS Reagent, Drum, 200 L
Reagents Acetone, ACS Reagent, Drum, 200 L
6228111

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Flash point: -20 °C (-4 °F)
  • Density: 0.8
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear Formula(CH3)2CO
  • ClassKetone
  • ChemistrySolvents
  • Synonyms2-Propanone, Dimethyl ketone
  • FormLiquid
  • Volume (mL)200000
  • Formula Weight58.08
  • Vapor Pressure (torr)176.7644
  • PackagingDrum
  • DOT InformationACETONE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number67-64-1
  • UN NumberUN1090
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)56.05
  • Boiling Point F (° F)132.89
  • Melting Point C (° C)-94.7
  • Melting Point F (° F)-138.46
  • Refractive Index1.3544
  • DescriptionAcetone, ACS Reagent, Drum Container, 200 L
Documents & Certificates
Reviews
Q&A