Reagents Acetic Acid, 10% (v/v), Standardized, Drum, 200 L

Representative image only.

Reagents Acetic Acid, 10% (v/v), Standardized, Drum, 200 L

Reagents | Mfr: CA008900-200E| Item: 6228883

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • 1:9
  • Assay 1.715 to 1.785 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1.01
  • Boiling point: approximately 118 °C (244.4 °F)
  • Melting poing: approximately 0 °C (32 °F)
  • Flash point: 39 °C (102.2 °F)
  • Solubility information: miscible
  • pH range: 2.3
  • 200 L drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Concentration10% v/v
  • ClassStandardized
  • FormLiquid
  • Volume (mL)200000
  • PackagingDrum
  • DOT InformationACETIC ACID SOLUTION, 8, P.G. III
  • Storage SymbolAmbient
  • Shelf Life (days)1095
  • Shelf Life (months)36
  • DescriptionAcetic Acid, 10% (v/v), Standardized, Drum, 200 L
Documents & Certificates
Reviews
Q&A