Reagents 2-Butoxyethanol, Amber Glass Bottle, 1 L

Representative image only.

Reagents 2-Butoxyethanol, Amber Glass Bottle, 1 L

Reagents | Mfr: C2156250-1C| Item: 6228446

Manufactured using the highest quality raw materials in ISO 9001 facility

Call For Price

Call For Price
Product Description

Key Features

  • Colorless liquid
  • Density: 0.9
  • Boiling point: 171 to 172 °C (339.8 to 341.6 °F)
  • Flash point: 62 °C (143.6 °F)
  • Refractive index: 1.418 to 1.42nD
  • Solubility information: miscible
  • 1 L amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Linear FormulaHOCH2CH2OC4H9
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsEthylene glycol monobutyl ether, Butoxyethanol(2-)
  • Volume (mL)1000
  • Formula Weight118.17
  • Vapor Pressure (torr)0..5925 - 09751
  • PackagingAmber glass bottle
  • DOT InformationTOXIC, LIQUIDS, ORGANIC, N.O.S. (ETHYLENE GLYCOL MONOBUTYL ETHER), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN2810
  • CAS Number111-76-2
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-70
  • Melting Point F (° F)-94
  • Description2-Butoxyethanol, Amber Glass Bottle, 1 L
Documents & Certificates
Reviews
Q&A