This item is now discontinued. Please browse similar items
Qsonica 4418 Ultrasonic Cell Distruptor Microtip Probe, 1/8" dia, 1 - 15 mL
Representative image only.
Qsonica 4418 Ultrasonic Cell Distruptor Microtip Probe, 1/8" dia, 1 - 15 mL
Qsonica | Mfr: 4418| Item: 0471262
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
Buy It Together
Current Item
Qsonica 4418 Ultrasonic Cell Distruptor Microtip Probe, 1/8" dia, 1 - 15 mL
Qsonica 4418 Ultrasonic Cell Distruptor Microtip Probe, 1/8" dia, 1 - 15 mL
0471262
£289.40GBP / Each
£347.28GBP (incl VAT)
+
Qsonica Q700 Sonicator® Touch-Screen Ultrasonic Processor; 700 watt, 120VAC, 50/60 Hz
0471282
£6,274.49GBP / Each
£7,529.39GBP (incl VAT)
+
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
£356.11GBP / Each
£427.33GBP (incl VAT)
=
Total Price 6920.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Microtips screw into the threaded end of standard ½" probe
  • 1 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.