Qsonica 4407 Replacement Tip for Standard Probes, 3/4" (19.1 mm)
Representative image only.
Qsonica 4407 Replacement Tip for Standard Probes, 3/4" (19.1 mm)
Qsonica | Mfr: 4407| Item: 0471273
Microtip Probes for use with Cole-Parmer and Qsonica Q500 and Q700 Sonicators
£74.67GBP / Each
£89.60GBP (incl VAT)
£74.67GBP / Each
£89.60GBP (incl VAT)
Buy It Together
Current Item
Qsonica 4407 Replacement Tip for Standard Probes, 3/4" (19.1 mm)
Qsonica 4407 Replacement Tip for Standard Probes, 3/4" (19.1 mm)
0471273
£74.67GBP / Each
£89.60GBP (incl VAT)
+
Qsonica Q700 Sonicator® Touch-Screen Ultrasonic Processor; 700 watt, 120VAC, 50/60 Hz
0471282
£6,274.49GBP / Each
£7,529.39GBP (incl VAT)
+
Cole-Parmer Coupler for 500- and 750-Watt Processors
0471085
£356.11GBP / Each
£427.33GBP (incl VAT)
=
Total Price 6705.27 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Microtips screw into the threaded end of standard ½" probe
  • 1 year warranty
More About this Item
Probes effectively process small samples and attach to the standard 1/2"-dia probe horn. The stepped microtip probe (04711-93) attaches directly to the converter and is ideal for reaching into long, thin tubes.