NSI Lab Solutions SS-10231-2 C. albicans Snap-Stick™ Swab; 2/PK
Representative image only.
NSI Lab Solutions SS-10231-2 C. albicans Snap-Stick™ Swab; 2/PK
NSI Lab Solutions | Item: 0500192
Easy-to-use Snap-Stick™ culture swab contains a pure microorganism
Call For Price
Call For Price
Product Description
Key Features
  • Hydration fluid dissolves culture pellet in less than a minute
  • Transparent tube ensures visual confirmation of pellet dissolution
  • Removable label prevents sample mix up
  • Glass-free culture swab device ensures user safety
  • No more than two passages from primary culture
  • No royalty fee
  • C. albicans NCPF 3179 culture
  • Pack of two
More About this Item
Use these Snap-Stick™ culture tubes containing microorganism pellets for qualitative microbiological analysis. To activate the microorganism, simply apply pressure to the plastic stem valve and release the hydration fluid to dissolve the microorganism pellet. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves in less than a minute with minimum agitation and no incubation. Transparent tube makes it easy to see if the pellet has completely dissolved. Once dissolved, remove the swab from the tube and swipe across the media plate. Each stick comes individually sealed and with a reusable label; use the label to mark your media plate to prevent sample mix up. Store refrigerated.
NCPF reference cultures obtained from the National Collection of Pathogenic Fungi.
Specifications
  • OrganismC. albicans
  • Packaging2/PK
  • Product TypeSnap-Stick™ Cultures
  • Shelf Life Months (months)24
  • Shipping requirementFrozen ice packs
  • Storage requirementRefrigerate between 2 and 8°C
  • StrainNCPF 3179, 10231
  • DescriptionC. albicans Snap-Stick™ Swab; 2/PK