NSI Lab Solutions SQCO-038B TOX in Soil PT Express™

Representative image only.

NSI Lab Solutions SQCO-038B TOX in Soil PT Express™

NSI Lab Solutions | Item: 6120235

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • One ready-to-use 100 g of sample
  • Designed for use with EPA Methods 9020B, 9065, 9066 and 9067
  • Contains Total Phenolics and TOX in the range of 0.5 to 100 mg/kg

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • Element/IonTotal Phenolics and TOX
  • Weight (g)100
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionTOX in Soil PT Express™
Documents & Certificates
Reviews
Q&A