NSI Lab Solutions SQCO-019B Nutrients in Soil PT Express™

Representative image only.

NSI Lab Solutions SQCO-019B Nutrients in Soil PT Express™

NSI Lab Solutions | Item: 6120357

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • One 40 g of sample for determination of Nutrients in the NELAC required range

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

For determination of:
Ammonia as N: 300 to 3000 mg/kg
Kjeldahl nitrogen (TKN): 400 to 4000 mg/kg
Total Phosphorus: 300 to 3000 mg/kg
Total Organic Carbon (TOC): 1000 to 15000 mg/kg

Specifications
  • Element/IonAmmonia as N, Kjeldahl nitrogen (TKN), Total Phosphorus, Total Organic Carbon (TOC)
  • FormSolid
  • Weight (g)40
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionNutrients in Soil PT Express™
Documents & Certificates
Reviews
Q&A