NSI Lab Solutions SQCO-005B PCB in Soil PT Express™; 2/PK

Representative image only.

NSI Lab Solutions SQCO-005B PCB in Soil PT Express™; 2/PK

NSI Lab Solutions | Item: 6120337

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • Supplied in duplicate for convenience and value
  • Two ready-to-use 30 g samples
  • Designed for use with EPA Method 8081
  • Contains one Arochlor per study
  • Formulated in the NELAC required range

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • Element/IonPCB
  • FormSolid
  • Weight (g)30
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8 °C
  • UN Number2315
  • Shelf Life (months)24
  • DescriptionPCB in Soil PT Express™; 2/PK
Documents & Certificates
Reviews
Q&A