NSI Lab Solutions SQCI-015B Anions in Soil PT Express™

Representative image only.

NSI Lab Solutions SQCI-015B Anions in Soil PT Express™

NSI Lab Solutions | Item: 6120294

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • One 40 g sample designed for the DI water extraction procedure followed by analyses for listed anions
  • Formulated in the NELAC required range where applicable.

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Analyze for the following anions:
Bromide: 10.0 to 100 mg/kg
Chloride: 200 to 1000 mg/kg
Fluoride: 25.0 to 500 mg/kg
Nitrate as N: 25.0 to 500 mg/kg
Nitrite as N: 25.0 to 500 mg/kg
Sulfate: 25.0 to 2000 mg/kg
Orthophosphate as P: 25.0 to 500 mg/kg
Nitrate+Nitrite as N: 50.0 to 1000 mg/kg

Specifications
  • Element/IonBromide, Chloride, Fluoride, Nitrate as N, Nitrite as N, Sulfate, Orthophosphate as P, Nitrate+Nitrite as N
  • FormSolid
  • Weight (g)40
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionAnions in Soil PT Express™
Documents & Certificates
Reviews
Q&A