NSI Lab Solutions SQCI-005 TCLP Metals in Soil Reference Material; 100 g Soil Blank and 21 mL Spiking Solution

Representative image only.

NSI Lab Solutions SQCI-005 TCLP Metals in Soil Reference Material; 100 g Soil Blank and 21 mL Spiking Solution

NSI Lab Solutions | Item: 6122632

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 1311, 1631, 200.7, 200.8, 206.2, 206.3, 208.1, 208.2, 210.1, 210.2, 213.1, 213.2, 245.1, 245.2, 270.2, 270.3, 272.1, 272.2, 279.1, 279.2, 286.1, 286.2, 289.1, 289.2, 6010, 6010B, 6020, 7020, 7040, 7041, 7060A, 7061A, 7062, 7080A, 7081, 7090, 7091, 7130, 7131A, 7190, 7191, 7200, 7201, 7210, 7211, 7380, 7381, 7420, 7421, 7460, 7461, 7470A, 7471, 7472, 7480, 7481, 7520, 7521, 7740, 7741A, 7742, 7760, 7760A, 7761, 7780, 7840, 7841, 7910, 7911, 7950, 7951, and SM 3120

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

Supplied as a 100 gram blank soil and a 21 mL spiking solution. Contains a subset of the metals listed.

Specifications
  • ConcentrationMultiple
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonMultiple
  • FormSolid
  • Volume (mL)21
  • Weight (g)100
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionTCLP Metals in Soil Reference Material; 100 g Soil Blank and 21 mL Spiking Solution
Documents & Certificates
Reviews
Q&A