NSI Lab Solutions SQCI-001B Metals in Soil PT Express™

Representative image only.

NSI Lab Solutions SQCI-001B Metals in Soil PT Express™

NSI Lab Solutions | Item: 6120295

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • One 40 g of ready-to-use sample in a bottle
  • Applicable to all ICP & AA - SW-846 and CLP Methods

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Contains all of the metals listed in the NELAC required range:
Aluminum: 2500 to 25,000 mg/kg
Antimony: 80.0 to 300 mg/kg
Arsenic: 40.0 to 400 mg/kg
Barium: 100 to 1000 mg/kg
Beryllium: 40.0 to 400 mg/kg
Boron: 80.0 to 800 mg/kg
Cadmium: 40.0 to 400 mg/kg
Calcium: 1500 to 25,000 mg/kg
Chromium: 40.0 to 400 mg/kg
Cobalt: 40.0v400 mg/kg
Copper: 40.0 to 400 mg/kg
Iron: 5000 to 50,000 mg/kg
Lead: 40.0 to 400 mg/kg
Lithium: 150 to 2000 mg/kg
Magnesium: 1200 to 25,000 mg/kg
Manganese: 100 to 2000 mg/kg
Mercury: 1.00 to 35.0 mg/kg
Molybdenum: 30.0 to 300 mg/kg
Nickel: 40.0 to 500 mg/kg
Potassium: 1400 to 25,000 mg/kg
Selenium: 40.0 to 400 mg/kg
Silver: 20.0 to 100 mg/kg
Sodium: 150 to 15000 mg/kg
Strontium: 40.0 to 400 mg/kg
Thallium: 40.0 to 400 mg/kg
Tin: 50.0 to 250 mg/kg
Titanium: 100 to 400 mg/kg
Vanadium: 40.0 to 400 mg/kg
Zinc: 100 to 1000 mg/kg

.

Specifications
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormSolid
  • Weight (g)40
  • Storage SymbolRefrigerate between 2 and 8 °C
  • Shelf Life (months)24
  • DescriptionMetals in Soil PT Express™
Documents & Certificates
Reviews
Q&A