NSI Lab Solutions SQC-113B PVOC in Soil PT Express™

Representative image only.

NSI Lab Solutions SQC-113B PVOC in Soil PT Express™

NSI Lab Solutions | Item: 6120258

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • A single ampule of concentrate in methanol and 15 g of clean soil matrix

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

For determination of:
Benzene: 1000 to 10,000 µg/kg
Ethylbenzene: 1000 to 10,000 µg/kg
Toluene: 1000 to 10,000 µg/kg
MTBE: 2000 to 10,000 µg/kg
Naphthalene: 2000 to 10,000 µg/kg
Total Xylenes: 2000 to 20,000 µg/kg
m+p-Xylene: 1000 to 10,000 µg/kg
o-Xylene: 1000 to 10,000 µg/kg

Specifications
  • Element/IonBenzene, Ethylbenzene, Toluene, MTBE, Naphthalene, Total Xylenes, m+p-Xylene, o-Xylene
  • FormLiquid
  • Storage SymbolAmpule: Refrigerate between 2 and 8 °CMatrix: Store between 15 to 30 °C
  • UN Number1230
  • Shelf Life (months)24
  • DescriptionPVOC in Soil PT Express™
Documents & Certificates
Reviews
Q&A