NSI Lab Solutions QCO-136B Nitroaromatics/Nitramines in Water Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-136B Nitroaromatics/Nitramines in Water Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120352

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in acetonitrile for determination of explosive residues in water

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample contains at least 80% of the following analytes formulated in the range of 1.0 to 20.0 µg/L.
1,3-Dinitrobenzene
1,3,5-Trinitrobenzene
2-Amino-4,6-dinitrotoluene
2-Nitrotoluene
2,4-Dinitrotoluene
2,4,6-Trinitrotoluene
2,6-Dinitrotoluene
3-Nitrotoluene
3,5 Dichloroaniline
4-Amino-2,6-dinitrotoluene
4-Nitrotoluene
HMX
Nitrobenzene
Nitroglycerin
Nitroguanidine
PETN
RDX
Tetryl

This sample is not listed in the TNI NPW Field of Testing.

Specifications
  • MatrixAcetonitile
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolStore between -20 and -10°C
  • UN Number1648
  • Shelf Life (months)24
  • DescriptionNitroaromatics/Nitramines in Water Wastewater PT Express™; 2/PK
Reviews
Q&A