NSI Lab Solutions QCO-121B Base/Neutrals Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-121B Base/Neutrals Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120376

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set for use with Methods 625/8270

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample design will satisfy PT requirements for any of the following analytes:
1,1-Biphenyl: 30 to 200 µg/L
1,2,4,5-Tetrachlorobenzene: 20 to 200 µg/L
1,2,4-Trichlorobenzene: 20 to 200 µg/L
1,2-Dichlorobenzene: 20 to 200 µg/L
1,2-Diphenylhydrazine: 30 to 200 µg/L
1,3,5-Trinitrobenzene: 20 to 200 µg/L
1,3-Dichlorobenzene: 20 to 200 µg/L
1,3-Dinitrobenzene: 20 to 200 µg/L
1,4-Dichlorobenzene: 20 to 200 µg/L
1,4-Dioxane: 20 to 200 µg/L
1,4-Naphthoquinone: 20 to 200 µg/L
1-Chloronaphthalene: 20 to 200 µg/L
1-Methylnaphthalene: 30 to 200 µg/L
1-Naphthylamine: 20 to 200 µg/L
2,3-Dichloroaniline: 20 to 200 µg/L
2,4-Dinitrotoluene: 20 to 200 µg/L
2,6-Dinitrotoluene: 20 to 200 µg/L
2-Acetylaminofluorene: 20 to 200 µg/L
2-Chloronaphthalene: 20 to 200 µg/L
2-Methylcholanthrene: 10 to 200 µg/L
2-Methylnaphthalene: 20 to 200 µg/L
2-Naphthylamine: 20 to 200 µg/L
2-Nitroaniline: 10 to 200 µg/L
2-Picoline: 20 to 200 µg/L
3,3-Dimethylbenzidine: 20 to 200 µg/L
3,3’-Dichlorobenzidine: 50 to 200 µg/L
3-Methylcholanthrene: 20 to 200 µg/L
3-Nitroaniline: 30 to 200 µg/L
4-Aminobiphenyl: 20 to 200 µg/L
4-Bromophenyl phenyl ether: 20 to 200 µg/L
4-Chloroaniline: 10 to 200 µg/L
4-Chlorophenyl phenyl ether: 20 to 200 µg/L
4-Nitroaniline: 10 to 200 µg/L
4-Nitroquineoline-1-oxide: 20 to 200 µg/L
5-Nitro-o-toluidine: 20 to 200 µg/L
7,12-Dimethylbenz(a)anthracene: 20 to 200 µg/L
a,a-Dimethylphenylamine: 20 to 200 µg/L
Acenaphthene: 10 to 200 µg/L
Acenaphthylene: 10 to 200 µg/L
Acetophenone: 20 to 200 µg/L
Aniline: 30 to 200 µg/L
Anthracene: 10 to 200 µg/L
Atrazine: 30 to 200 µg/L
Benzaldehyde: 30 to 200 µg/L
Benzidine: 200 to 1000 µg/L
Benzo(a)anthracene: 10 to 200 µg/L
Benzo(a)pyrene: 10 to 200 µg/L
Benzo(b)fluoranthene: 20 to 200 µg/L
Benzo(g,h,i)perylene: 10 to 200 µg/L
Benzo(k)fluoranthene: 20 to 200 µg/L
Benzyl alcohol: 30 to 200 µg/L
Benzyl butyl phthalate: 50 to 200 µg/L
bis(2-Chloroethoxy)methane: 20 to 200 µg/L
bis(2-Chloroethyl)ether: 20 to 200 µg/L
2,2’-Oxybis(1-Chloropropane): 30 to 200 µg/L
bis(2-Ethylhexyl)phthalate: 20 to 200 µg/L
Caprolactam: 30 to 200 µg/L
Carbazole: 20 to 200 µg/L
Chlorobenzilate: 20 to 200 µg/L
Chrysene: 10 to 200 µg/L
Di-n-butyl phthalate: 40 to 200 µg/L
Di-n-octyl phthalate: 30 to 200 µg/L
Diallate: 20 to 200 µg/L
Dibenz(a,h)anthracene: 20 to 200 µg/L
Dibenzofuran: 30 to 200 µg/L
Diethyl phthalate: 50 to 200 µg/L
Dimethoate: 20 to 200 µg/L
Dimethyl phthalate: 50 to 200 µg/L
Dinoseb: 20 to 200 µg/L
Diphenyl ether: 20 to 200 µg/L
Disulfoton: 20 to 200 µg/L
Ethyl methanesulfonate : 30 to 200 µg/L
Famphur: 20 to 200 µg/L
Fluoranthene: 30 to 200 µg/L
Fluorene: 10 to 200 µg/L
Hexachlorobenzene: 20 to 200 µg/L
Hexachlorobutadiene: 50 to 200 µg/L
Hexachlorocyclopentadiene: 50 to 200 µg/L
Hexachloroethane: 50 to 200 µg/L
Hexachlorophene: 20 to 200 µg/L
Hexachloropropene: 20 to 200 µg/L
Indeno(1,2,3-c,d)pyrene: 30 to 200 µg/L
Isodrin: 20 to 200 µg/L
Isophorone: 20 to 200 µg/L
Isosafrole: 20 to 200 µg/L
Kepone: 20 to 200 µg/L
m-Dinitrobenzene: 10 to 200 µg/L
Methapyrilene: 20 to 200 µg/L
Methyl methanesulfonate: 10 to 200 µg/L
Methyl parathion: 20 to 200 µg/L
n-Decane: 20 to 200 µg/L
N-Nitroso-di-n-butylamine: 20 to 200 µg/L
N-Nitroso-di-n-propylamine: 30 to 200 µg/L
N-Nitrosodiethylamine: 20 to 200 µg/L
N-Nitrosodimethylamine: 75 to 200 µg/L
N-Nitrosodiphenylamine: 30 to 200 µg/L
N-Nitrosomorpholine: 20 to 200 µg/L
N-Nitrosopiperidine: 20 to 200 µg/L
N-Nitrosopyrrolidine: 20 to 200 µg/L
n-Octadecane: 20 to 200 µg/L
Naphthalene: 20 to 200 µg/L
Nitrobenzene: 20 to 200 µg/L
o,o,o-Triethylphoshorothioate: 20 to 200 µg/L
o-Dinitrobenzene: 10 to 200 µg/L
o-Toluidine: 20 to 200 µg/L
p-Dimethylaminoazobenzene: 20 to 200 µg/L
p-Dinitrobenzene: 10 to 200 µg/L
p-Phenylenediamine: 20 to 200 µg/L
Parathion: 20 to 200 µg/L
Pentachlorobenzene: 20 to 200 µg/L
Pentachlorohexane: 20 to 200 µg/L
Pentachloronitrobenzene: 20 to 200 µg/L
Phenacetin: 20 to 200 µg/L
Phenanthrene: 10 to 200 µg/L
Phorate: 20 to 200 µg/L
Pronamide: 20 to 200 µg/L
Pyrene: 10 to 200 µg/L
Pyridine: 10 to 200 µg/L
Safrole: 20 to 200 µg/L
Sulfotepp: 20 to 200 µg/L
Thionazin: 20 to 200 µg/L

Specifications
  • MatrixAcetone/Methanol solution
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1090/1230
  • Shelf Life (months)24
  • DescriptionBase/Neutrals Wastewater PT Express™; 2/PK
Reviews
Q&A