NSI Lab Solutions QCO-120B Volatiles Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-120B Volatiles Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120304

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in methanol for use with Methods 601/602, 8010/8020, 624, 8240, and 8260

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample design will satisfy PT requirements for any of the following analytes:
1,1-Dichloroethane: 10 to 150 µg/L
1,1-Dichloroethene: 10 to 150 µg/L
1,1,1-Trichloroethane: 10 to 100 µg/L
1,1,1,2-Tetrachloroethane: 15 to 150 µg/L
1,1,2-Trichloroethane: 15 to 150 µg/L
1,1,2,2-Tetrachloroethane: 15 to 150 µg/L
1,2-Dibromo-3-chloropropane: 15 to 150 µg/L
1,2-Dichlorobenzene: 10 to 120 µg/L
1,2-Dichloroethane: 15 to 150 µg/L
1,2-Dichloropropane: 10 to 150 µg/L
1,2,3-Trichlorobenzene: 15 to 150 µg/L
1,2,3-Trichloropropane: 15 to 150 µg/L
1,2,4-Trichlorobenzene: 15 to 150 µg/L
1,2,4-Trimethylbenzene: 10 to 120 µg/L
1,3,5-Trimethylbenzene: 10 to 120 µg/L
1,3-Dichlorobenzene: 10 to 120 µg/L
1,4-Dichlorobenzene: 10 to 120 µg/L
1,4-Dioxane: 20 to 500 µg/L
2-Butanone: 5 to 500 µg/L
2-Chloroethyl vinyl ether: 5 to 500 µg/L
2-Hexanone: 20 to 200 µg/L
4-Methyl-2-pentanone: 20 to 200 µg/L
Acetone: 20 to 200 µg/L
Acetonitrile: 5 to 500 µg/L
Acrolein: 5 to 500 µg/L
Acrylonitrile: 5 to 500 µg/L
Benzene: 10 to 120 µg/L
Bromodichloromethane: 10 to 100 µg/L
Bromoform: 10 to 100 µg/L
Bromomethane: 20 to 120 µg/L
Carbon disulfide: 5 to 500 µg/L
Carbon tetrachloride: 15 to 150 µg/L
Chlorobenzene: 10 to 120 µg/L
Chloroethane: 20 to 120 µg/L
Chloroform: 10 to 100 µg/L
Chloromethane: 20 to 120 µg/L
cis-1,2-Dichloroethene: 10 to 150 µg/L
cis-1,3-Dichloropropene: 10 to 120 µg/L
Dibromochloromethane: 10 to 100 µg/L
Dibromomethane: 10 to 120 µg/L
Dichlorodifluoromethane: 20 to 100 µg/L
Ethylbenzene: 10 to 120 µg/L
Ethylene dibromide: 10 to 120 µg/L
Methyl acetate: 5 to 500 µg/L
Methyl cyclohexane: 20 to 100 µg/L
Methylene chloride: 10 to 120 µg/L
m+p-Xylene: 10 to 150 µg/L
MTBE: 15 to 150 µg/L
Naphthalene: 15 to 150 µg/L
n-Hexane: 10 to 150 µg/L
o-Xylene: 10 to 150 µg/L
Styrene: 20 to 120 µg/L
Tetrachloroethene: 10 to 150 µg/L
Toluene: 10 to 120 µg/L
Total Xylenes: 20 to 300 µg/L
trans-1,2-Dichloroethene: 10 to 120 µg/L
trans-1,3-Dichloropropene: 10 to 120 µg/L
Trichloroethene: 10 to 100 µg/L
Trichlorofluoromethane: 20 to 120 µg/L
Vinyl acetate: 5 to 500 µg/L
Vinyl chloride: 20 to 120 µg/L

Specifications
  • MatrixMethanol solution
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1230
  • Shelf Life (months)24
  • DescriptionVolatiles Wastewater PT Express™; 2/PK
Reviews
Q&A