NSI Lab Solutions QCO-022B Acids Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-022B Acids Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120219

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in acetone for use with Methods 604/8040/8041 or 625/8270

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample design will satisfy PT requirements for any of the following analytes:
2-Chlorophenol: 30 to 200 µg/L
2-Cyclohexyl-4,6-dinitrophenol: 50 to 200 µg/L
2-Methyl-4,6-dinitrophenol: 40 to 200 µg/L
2-Methylphenol: 40 to 200 µg/L
2-Nitrophenol: 50 to 200 µg/L
2,3,4,5-Tetrachlorophenol: 50 to 200 µg/L
2,3,4,6-Tetrachlorophenol: 50 to 200 µg/L
2,4-Dichlorophenol: 30 to 200 µg/L
2,4-Dimethylphenol: 40 to 200 µg/L
2,4-Dinitrophenol: 100 to 200 µg/L
2,4,5-Trichlorophenol: 30 to 200 µg/L
2,4,6-Trichlorophenol: 30 to 200 µg/L
2,6-Dichlorophenol: 30 to 200 µg/L
4-Chloro-3-methylphenol: 30 to 200 µg/L
4-Methylphenol: 50 to 200 µg/L
4-Nitrophenol: 100 to 200 µg/L
Benzoic acid: 50 to 200 µg/L
Pentachlorophenol: 40 to 200 µg/L
Phenol: 100 to 200 µg/L

Specifications
  • MatrixAcetone
  • Element/IonMultiple (see the full listing under "More About This Item")
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1090
  • Shelf Life (months)24
  • DescriptionAcids Wastewater PT Express™; 2/PK
Reviews
Q&A