NSI Lab Solutions QCO-020B PCB in Water Wastewater PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-020B PCB in Water Wastewater PT Express™; 2/PK

NSI Lab Solutions | Item: 6120361

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Submit PT report to accreditation agencies at no additional charge
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in acetone for use with Methods 608/8080/8081

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

For determination of the following elements:
Arochlor 1016: 2.0 to 10 µg/L
Arochlor 1221: 2.0 to 10 µg/L
Arochlor 1232: 2.0 to 10 µg/L
Arochlor 1242: 2.0 to 10 µg/L
Arochlor 1248: 2.0 to 10 µg/L
Arochlor 1254: 2.0 to 10 µg/L
Arochlor 1260: 2.0 to 10 µg/L
Arochlor 1262: 2.0 to 10 µg/L
Arochlor 1268: 2.0 to 10 µg/L

Specifications
  • MatrixAcetone
  • Element/IonArochlor 1016, Arochlor 1221, Arochlor 1232, Arochlor 1242, Arochlor 1248, Arochlor 1254, Arochlor 1260, Arochlor 1268
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1090
  • Shelf Life (months)24
  • DescriptionPCB in Water Wastewater PT Express™; 2/PK
Reviews
Q&A