NSI Lab Solutions QCO-020 PCB in Water QC Concentrate; 1.5 mL

Representative image only.

NSI Lab Solutions QCO-020 PCB in Water QC Concentrate; 1.5 mL

NSI Lab Solutions | Item: 6122615

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 608, 8080, and 8082

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 1.5 mL concentrate in Acetone for use with Methods 608/8080/8081.

Specifications
  • Concentration2 to 10 µg/L
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonAroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260
  • FormLiquid
  • Volume (mL)1.5
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionPCB in Water QC Concentrate; 1.5 mL
Documents & Certificates
Reviews
Q&A