NSI Lab Solutions QCO-006 Regulated SOCs QC Concentrate; 1.5 mL

Representative image only.

NSI Lab Solutions QCO-006 Regulated SOCs QC Concentrate; 1.5 mL

NSI Lab Solutions | Item: 6122611

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 506, 525, and 550

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 1.5 mL concentrate in Acetone for use with Methods 506/525/550. Each sample includes Benzo(a)pyrene - 0.2 - 2.5ug/L, bis(2-Ethylhexyl)phthalate - 5 - 50 ug/L, bis(2-Ethylhexyl)adipate - 8 - 50 ug/L, plus a subset of analytes drawn from the following list:

Specifications
  • ConcentrationMultiple
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1.5
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionRegulated SOCs QC Concentrate; 1.5 mL
Documents & Certificates
Reviews
Q&A