NSI Lab Solutions QCO-001B Carbamate Pesticides Drinking Water PT Express™; 2/PK

Representative image only.

NSI Lab Solutions QCO-001B Carbamate Pesticides Drinking Water PT Express™; 2/PK

NSI Lab Solutions | Item: 6120464

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in methanol for use with Method 531.1

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry drinking water proficiency tests are designed for identification of target chemicals in drinking water.

Environmental Chemistry Drinking Water PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

The sample design will satisfy PT reqµirements for the following analytes:
Aldicarb: 15 to 100 µg/L
Aldicarb sulfone: 15 to 100 µg/L
Aldicarb sulfoxide: 15 to 80 µg/L
Baygon: 30 to 140 µg/L
Carbaryl: 15 to 100 µg/L
Carbofuran: 15 to 150 µg/L
3-Hydroxy carbofuran: 15 to 80 µg/L
Methiocarb: 30 to 140 µg/L
Methomyl: 15 to 100 µg/L
Oxamyl (Vydate): 15 to 100 µg/L

Specifications
  • MatrixMethanol
  • Element/IonAldicarb, Aldicarb sulfone, Aldicarb sulfoxide, Baygon, Carbaryl, Carbofuran, 3-Hydroxy carbofuran, Methiocarb, Methomyl, Oxamyl (Vydate)
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1230
  • Shelf Life (months)24
  • DescriptionCarbamate Pesticides Drinking Water PT Express™; 2/PK
Reviews
Q&A