NSI Lab Solutions QCO-001 Carbamate Pesticides (WS) QC Concentrate; 1.5 mL

Representative image only.

NSI Lab Solutions QCO-001 Carbamate Pesticides (WS) QC Concentrate; 1.5 mL

NSI Lab Solutions | Item: 6122605

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • Aldicarb 15-100 ug/L
  • Oxamyl (Vydate) 15-100 ug/L
  • Aldicarb sulfone 15-100 ug/L
  • Baygon 30-140 ug/L
  • Aldicarb sulfoxide 15-80 ug/L
  • Carbaryl 15-100 ug/L
  • Carbofuran 15-150 ug/L
  • Methomyl 15-100 ug/L
  • Methiocarb 30-140 ug/L
  • 3-Hydroxy carbofuran 15-80 ug/L

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 1.5 mL concentrate in Methanol for use with Method 531.1. Analytes and possible range listed below. Sample designed to be diluted 1mL to 1000mL in reagent water. Derived from previous PT study. Useful for method validation and analyst DOCs.

Specifications
  • ClassQC Samples
  • ChemistryOrganics;Standards
  • FormLiquid
  • Volume (mL)1.5
  • DescriptionCarbamate Pesticides (WS) QC Concentrate; 1.5 mL
Documents & Certificates
Reviews
Q&A