NSI Lab Solutions QCI-156C Turbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 4/Pk

Representative image only.

NSI Lab Solutions QCI-156C Turbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 4/Pk

NSI Lab Solutions | Item: 0500509

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • A formazin based CRM
  • 5 NTU
  • Pack of four 500 mL bottles

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration5 NTU
  • MatrixWater
  • ClassTurbidity QC CRM
  • ChemistryInorganics;Standards
  • Element/IonTurbidity
  • FormLiquid
  • Volume (mL)500
  • Packaging4/PK
  • DOT InformationNot Regulated by DOT
  • Storage SymbolStore between 15 and 30 °C
  • Hazard SymbolIrritant
  • Shelf Life (months)24
  • DescriptionTurbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 4/PK
Documents & Certificates
Reviews
Q&A