NSI Lab Solutions QCI-156 Turbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 1/EA

Representative image only.

NSI Lab Solutions QCI-156 Turbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 1/EA

NSI Lab Solutions | Item: 0500508

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • A formazin based CRM
  • 5 NTU
  • One 500 mL bottle

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration5 NTU
  • MatrixWater
  • ClassTurbidity QC CRM
  • ChemistryInorganics;Standards
  • Element/IonTurbidity
  • FormLiquid
  • Volume (mL)500
  • Packaging1/EA
  • DOT InformationNot Regulated by DOT
  • Storage SymbolStore between 15 and 30 °C
  • Hazard SymbolIrritant
  • Shelf Life (months)24
  • DescriptionTurbidity QC Check Certified Reference Material, 5 NTU, 500 mL; 1/EA
Documents & Certificates
Reviews
Q&A