NSI Lab Solutions QCI-147S Stable Sulfide Certified Reference Material, 10 mg/mL, 2 mL; 1/EA

Representative image only.

NSI Lab Solutions QCI-147S Stable Sulfide Certified Reference Material, 10 mg/mL, 2 mL; 1/EA

NSI Lab Solutions | Item: 0500507

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Supplied ready to use—no lengthy preparative steps required
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Precipitate-free preservation using a proprietary buffer system and inert gas sealing
  • 10 mg/mL sulfide
  • One 2 mL ampule

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration10 mg/mL
  • Element/IonSulfide (from Na2S)
  • FormLiquid
  • Volume (mL)2
  • Packaging1/EA
  • Storage SymbolRefrigerate between 2 and 8 °C
  • CAS Number1313-82-2
  • UN Number1824
  • Shelf Life (months)24
  • DescriptionStable Sulfide Certified Reference Material, 10 mg/mL, 2 mL; 1/EA
Documents & Certificates
Reviews
Q&A