NSI Lab Solutions QCI-137 Hardness QC Certified Reference Material; 250 mL

Representative image only.

NSI Lab Solutions QCI-137 Hardness QC Certified Reference Material; 250 mL

NSI Lab Solutions | Item: 0500547

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 130.1, 130.2, 160.2, 200.7, 200.8, 215.1, 215.2, 242.1, 6010, 6010B, 6020, 7140, 7450; SM 18/19th ED 2340 B; SM 18/19th ED 2340 C; SM 18/19th ED 3500 Ca-D; SM 18/19th ED 3500-Mg E; SM 18th ED 2540 D; SM 18th ED 3500-Ca D; SM 18th ED 3500-Mg D; and SM 3120

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 250 mL ready-to-use sample packaged in a HDPE bottle to be analyzed for:  Calcium 10 - 100 mg/L, Magnesium 4.0 - 40 mg/L, Total Hardness 40 - 415 mg/L, Calcium Hardness 25 - 250 mg/L. Exact values are documented on lot COA.

Specifications
  • ConcentrationMultiple
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)250
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionHardness QC Certified Reference Material; 250 mL
Documents & Certificates
Reviews
Q&A