NSI Lab Solutions QCI-129 SGT - HEM (TPH) QC Concentrate, Certified Reference Material; 5.0 mL

Representative image only.

NSI Lab Solutions QCI-129 SGT - HEM (TPH) QC Concentrate, Certified Reference Material; 5.0 mL

NSI Lab Solutions | Item: 6122599

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 5.0 mL sample for dilution to 1000 mL. Can be used for IR methods as well as Gravimetric methods. Formulated in the NELAC range of 20 - 200 mg/L. Each ampule produces 5 liters of sample.

Specifications
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • FormLiquid
  • Volume (mL)5
  • DescriptionSGT - HEM (TPH) QC Concentrate, Certified Reference Material; 5.0 mL
Documents & Certificates
Reviews
Q&A