NSI Lab Solutions QCI-127 Volatile Solids QC Concentrate, Certified Reference Material

Representative image only.

NSI Lab Solutions QCI-127 Volatile Solids QC Concentrate, Certified Reference Material

NSI Lab Solutions | Item: 6122597

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Method 160.4

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A screw-cap vial containing a solid material for dilution to 1000 mL. Formulated in the NELAC range of 100 - 500 mg/L. Each vial produces at least 1 liter of sample.

Specifications
  • Concentration100 to 500 µg/mL
  • ClassQC Samples
  • ChemistryStandards;Inorganics
  • Element/IonVolatile Solids, Total
  • FormLiquid
  • Volume (mL)12
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionVolatile Solids QC Concentrate, Certified Reference Material
Documents & Certificates
Reviews
Q&A