NSI Lab Solutions QCI-126 Settleable Solids QC Standard, Certified Reference Material
Representative image only.
NSI Lab Solutions QCI-126 Settleable Solids QC Standard, Certified Reference Material
NSI Lab Solutions | Item: 6122596
Meet or exceed performance criteria with QC samples
Call For Price
Call For Price
Product Description
Key Features
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
More About this Item
The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.
A natural solid for quantitative transfer to a 1 liter Class A volumetric flask with dilution to 1 liter in reagent water. Formulated in the NELAC range of 5.0 - 50 mL/L. Each vial produces 1 liter of sample.
Specifications
  • ClassQC Samples
  • ChemistryStandards;Inorganics
  • FormSolid
  • DescriptionSettleable Solids QC Standard, Certified Reference Material