NSI Lab Solutions QCI-099 Acidity QC Concentrate, Certified Reference Material; 125 mL

Representative image only.

NSI Lab Solutions QCI-099 Acidity QC Concentrate, Certified Reference Material; 125 mL

NSI Lab Solutions | Item: 6122591

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Method 305.1 and SM 18th ED 2310 B (4A)

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 125 mL sample for determination of Acidity. Formulated in the NELAC range of 650 - 1800 mg/L.  Sample derived from our accredited inter-laboratory PT studies.  Study statistics available.  We always have 4 lots available so please call us at 1-800-234-7837 if you have a specific need.

Specifications
  • Concentration650 to 1,800 µg/mL
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonAcidity as CaCO3
  • FormLiquid
  • Volume (mL)125
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionAcidity QC Concentrate, Certified Reference Material; 125 mL
Documents & Certificates
Reviews
Q&A