NSI Lab Solutions QCI-079 Residue QC Concentrate, Certified Reference Material; 500 mL

Representative image only.

NSI Lab Solutions QCI-079 Residue QC Concentrate, Certified Reference Material; 500 mL

NSI Lab Solutions | Item: 6122585

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 160.1, 160.2, 160.3; and SM 18/19th ED 2540 C; SM 18th ED 2540 B; and SM 18th ED 2540 D

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 500 mL ready-to-use whole volume sample to be analyzed for Total Suspended Solids in the NELAC range of 20-100 mg/L, Total Dissolved Solids in the NELAC range of 140-800 mg/L, and Total Solids formulated in the NELAC range of 140-800 mg/L.

Specifications
  • ConcentrationMultiple
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionResidue QC Concentrate, Certified Reference Material; 500 mL
Documents & Certificates
Reviews
Q&A