NSI Lab Solutions QCI-073 Silica Drinking Water QC Concentrate, Certified Reference Material; 21 mL

Representative image only.

NSI Lab Solutions QCI-073 Silica Drinking Water QC Concentrate, Certified Reference Material; 21 mL

NSI Lab Solutions | Item: 6122584

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 200.7, 200.8, 6010; SM 18/19th ED 3111 D; SM 18/19th ED 3120 B; SM 18/19th ED 4500-Si-E; and SM 18th ED 4500-Si D

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 21 mL concentrate for determination of Silica. Formulated in the NELAC range of 5 - 75 mg/L. Each vial produces 2 liters of sample.

Specifications
  • Concentration5 to 75 µg/mL
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonSilica as SiO2
  • FormLiquid
  • Volume (mL)21
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionSilica Drinking Water QC Concentrate, Certified Reference Material; 21 mL
Documents & Certificates
Reviews
Q&A