NSI Lab Solutions QCI-069 Oil & Grease Ready-To-Use Certified Reference Material, 1 L; 1/EA

Representative image only.

NSI Lab Solutions QCI-069 Oil & Grease Ready-To-Use Certified Reference Material, 1 L; 1/EA

NSI Lab Solutions | Item: 0500496

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Supplied ready to use—no lengthy preparative steps required
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • QC Standard to QA all of your total oil and grease analyses
  • Supplied ready-to-use—no lengthy preparative steps required
  • Formulated in HCL preserved reagent water
  • Entire contents must be analyzed for accurate results
  • Works with hexane or freon extractions
  • A NIST Traceable Reference Material
  • One 1000 mL amber bottle

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration129 mg/L
  • Element/IonOil, Grease
  • FormLiquid
  • Volume (mL)1000
  • Packaging1/EA
  • Storage SymbolStore between 15 and 30 °C
  • UN Number1789
  • CAS Number7647-01-0, 71-23-8, 8001-22-7, 8042-47-5
  • Shelf Life (months)24
  • DescriptionOil & Grease Ready-To-Use Certified Reference Material, 1 L; 1/EA
Documents & Certificates
Reviews
Q&A