NSI Lab Solutions QCI-064 Complex Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA

Representative image only.

NSI Lab Solutions QCI-064 Complex Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA

NSI Lab Solutions | Item: 0500543

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • For determination of TKN and Total Phosphorus
  • One 500 mL bottle

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Ready-to-use bottle contains:
TKN: 3.0 to 35 mg/L
Total Phosphorus: 0.5 to 10 mg/L

Specifications
  • MatrixWater
  • ClassComplex Nutrients QC CRM
  • ChemistryInorganics;Standards
  • Element/IonTKN, Total Phosphorus
  • FormLiquid
  • Volume (mL)500
  • Packaging1/EA
  • DOT InformationNot Regulated by DOT
  • Storage SymbolStore between 2 and 8 °C
  • Hazard SymbolIrritant
  • CAS Number56-40-6, 13408-09-8
  • Shelf Life (months)24
  • DescriptionComplex Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA
Documents & Certificates
Reviews
Q&A