NSI Lab Solutions QCI-063 Simple Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA

Representative image only.

NSI Lab Solutions QCI-063 Simple Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA

NSI Lab Solutions | Item: 0500541

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • For determination of Ammonia as N, Orthophosphate as P, Nitrate as N, and Nitrate/Nitrite-N
  • One 500 mL bottle

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Ready-to-use bottle contains:
Ammonia as N: 1.0 to 20 mg/L
Orthophosphate as P: 0.5 to 5.5 mg/L
Nitrate as N: 2.0 to 25 mg/L
Nitrate/Nitrite-N: 2.5 to 25 mg/L

Specifications
  • MatrixWater
  • ClassSimple Nutrients QC CRM
  • ChemistryStandards;Inorganics
  • Element/IonAmmonia as N, Orthophosphate as P, Nitrate as N, Nitrate/Nitrite-N
  • FormLiquid
  • Volume (mL)500
  • Packaging1/EA
  • DOT InformationNot Regulated by DOT
  • Storage SymbolStore between 2 and 8 °C
  • Hazard SymbolIrritant
  • CAS Number12125-02-9, 7631-99-4, 7758-11-4
  • Shelf Life (months)24
  • DescriptionSimple Nutrients QC Concentrate Certified Reference Material, 500 mL; 1/EA
Documents & Certificates
Reviews
Q&A