NSI Lab Solutions QCI-055H High-Level Solids Certified Reference Material, 1 L; 4/PK

Representative image only.

NSI Lab Solutions QCI-055H High-Level Solids Certified Reference Material, 1 L; 4/PK

NSI Lab Solutions | Item: 0500494

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Supplied ready-to-use—just shake and pour!
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • For high-level determination of TSS, TDS, and TS
  • Actual concentrations may vary slightly lot to lot
  • Certified stable for up to 2 years from manufacture
  • A NIST Traceable Certified Reference Material

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Ready-to-use bottle contains:
Total Dissolved Solids: 3000 mg/L
Total Solids: 3500 mg/L
Total Suspended Solids: 500 mg/L

Specifications
  • Element/IonTotal Suspended Solids, Total Dissolved Solids, Total Solids
  • FormLiquid
  • Volume (mL)1000
  • Packaging4/PK
  • Storage SymbolStore between 15 and 30°C
  • Shelf Life (months)24
  • DescriptionHigh-Level Solids Certified Reference Material, 1 L; 4/PK
Documents & Certificates
Reviews
Q&A