NSI Lab Solutions QCI-054 LAS (MBAs, Anionic Surfactant) Certified Reference Material, 1000 mg/L, 10 mL; 1/EA

Representative image only.

NSI Lab Solutions QCI-054 LAS (MBAs, Anionic Surfactant) Certified Reference Material, 1000 mg/L, 10 mL; 1/EA

NSI Lab Solutions | Item: 0500492

Validate analytical methods and evaluate the quality of your results with certified reference materials

Call For Price

Call For Price
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Certified stable for up to 2 years from manufacture
  • Supplied ready to use—no lengthy preparative steps required
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Flame sealed amber ampule
  • Source is Dodecene-1 sulfonate
  • A NIST Traceable Reference Material

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Specifications
  • Concentration1000 mg/L
  • Element/IonDodecene-1 LAS
  • FormLiquid
  • Volume (mL)10
  • Packaging1/EA
  • Storage SymbolStore between 15 and 30 °C
  • CAS Number25155-30-0
  • Shelf Life (months)24
  • DescriptionLAS (MBAs, Anionic Surfactant) Certified Reference Material, 1000 mg/L, 10 mL; 1/EA
Documents & Certificates
Reviews
Q&A