NSI Lab Solutions QCI-041 Inorganics QC Certified Reference Material; 500 mL

Representative image only.

NSI Lab Solutions QCI-041 Inorganics QC Certified Reference Material; 500 mL

NSI Lab Solutions | Item: 0500539

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Methods 200.7, 200.8, 258.1, 273.1, 273.2, 300.0, 325.1, 325.2, 325.3, 340.1, 340.2, 340.3, 6010, 6010B, 6020, 9056, 9212, 9214, 9250, 9251, 9253; SM 18/19th ED 4500 Cl- B; SM 18/19th ED 4500 Cl- D; SM 18/19th ED 4500 F-B; SM 18/19th ED 4500 F-C; SM 18/19th ED 4500 F-D; SM 18/19th ED 4500 F-E; SM 18/19th ED 4500-Cl E; SM 18/19th ED 4500-Cl F; SM 18th ED 3500 Na D; SM 18th ED 3500-K D; SM 18th ED 4500 Cl B; SM 18th ED 4500 Cl C; SM 18th ED 4500 Cl- E; SM 18th ED 4500-Cl D; SM 18th ED 4500-Cl F; and SM 3120

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 500 mL ready-to-use sample packaged in a HDPE bottle for determination of Potassium 10 - 40 mg/L, Nitrate-N 3 - 10 mg/L, Nitrate/Nitrite-N 3 - 10 mg/L, Chloride 20 - 160 mg/L, Sulfate 25 - 250 mg/L, Fluoride 1 - 8 mg/L, TDS at 180°C 100 - 1000 mg/L, Conductivity 130 - 1300 umhos/cm, Alkalinity 25 - 200 mg/L.

Specifications
  • ConcentrationMultiple
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionInorganics QC Certified Reference Material; 500 mL
Documents & Certificates
Reviews
Q&A