NSI Lab Solutions QCI-014 Turbidity Drinking Water QC Concentrate, Certified Reference Material; 21 mL

Representative image only.

NSI Lab Solutions QCI-014 Turbidity Drinking Water QC Concentrate, Certified Reference Material; 21 mL

NSI Lab Solutions | Item: 6122573

Meet or exceed performance criteria with QC samples

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for AA, ICP, ICP-MS, IC, XRF, and other analytical instrumentation
  • For US EPA Method 180.1 and SM 18/19th ED 2130 B

More About this Item

The end-product of most analytical laboratories is data. It is therefore imperative that the quality of the data produced meet or exceed performance criteria. The implementation of Quality Control (QC) processes helps control error and estimate uncertainty. QC can be a tool in method development, regulatory processes, laboratory training, and performance monitoring. QC samples can help an analytical laboratory determine its precision, accuracy, linear range, and method ruggedness. QC samples allow the laboratories to fulfill documentation requirements as well as correct errors in root case analysis of a possible failure. Finally, QC samples monitor the effectiveness of the laboratory analysts to produce accurate data.

A 21 mL concentrate for determination of Turbidity in the NELAC range of 0.5 - 8 NTU. Formazin based. Each container produces 2 liters of sample. Sample derived from interlab PT program.

Specifications
  • Concentration0.5 to 8 NTU
  • ClassQC Samples
  • ChemistryInorganics;Standards
  • Element/IonTurbidity
  • FormLiquid
  • Volume (mL)21
  • DOT InformationNon-Hazardous
  • Storage SymbolAmbient
  • DescriptionTurbidity Drinking Water QC Concentrate, Certified Reference Material; 21 mL
Documents & Certificates
Reviews
Q&A