NSI Lab Solutions QC-116B TPH in Water PT Express™

Representative image only.

NSI Lab Solutions QC-116B TPH in Water PT Express™

NSI Lab Solutions | Item: 6120259

On-demand proficiency test (PT) for when you need results fast

Call For Price

Call For Price
Product Description

Key Features

  • Fast turnaround—PT reports available within 48 hours!
  • Available year-round
  • Convenient online reporting with PT Datalink
  • Submit PT report to accreditation agencies at no additional charge
  • ISO 17043 accredited
  • A sample concentrate for analysis of TPH in water by IR or Gravimetric Methods
  • Single ampule

More About this Item

The proficiency testing studies help improve quality and comply with regulations. Order the NSI PT Express™, our on-demand proficiency testing, when you have an immediate need for proficiency test results.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

Environmental Chemistry Wastewater PT Express samples may take 7 to 10 business days to ship. If you need the samples sooner, please call NSI at 1-800-234-7837 to expedite your order.

Specifications
  • Element/IonTPH
  • FormLiquid
  • Storage SymbolStore between 15 to 30 °C
  • UN Number1090
  • Shelf Life (months)24
  • DescriptionTPH in Water PT Express™
Documents & Certificates
Reviews
Q&A