NSI Lab Solutions QC-003LSNIP Oil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK

Representative image only.

NSI Lab Solutions QC-003LSNIP Oil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK

NSI Lab Solutions | Item: 0500519

Validate analytical methods and evaluate the quality of your results with premeasured certified reference materials

Call For Price

Call For Price

Buy It Together

Current Item

NSI Lab Solutions QC-003LSNIP Oil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK
NSI Lab Solutions QC-003LSNIP Oil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK
0500519

Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Manufactured from highest purity water and analytically verified source materials
  • Standards are traceable to SI through NIST test masses and/or SRMs
  • Manufactured and certified under ISO 17034 accreditation
  • Meets your ISO 17025 requirements
  • Convenient, easy to use and accurate single-use tube
  • For Oil and Grease Method 1664
  • Certified stable for up to 2 years from manufacture
  • 0.2% concentration
  • 20 x 10 mL Snips per pack

More About this Item

Accuracy, homogeneity, and performance of each lot is certified in our ISO 17025 accredited laboratory by the same methods you use.

Using a Snip is a Snap! Each tube delivers exactly 40 mg of total oil and grease. To use, snip open the PTFE tube and pour contents directly into extraction sample. There is no need to pipette.

Specifications
  • Concentration0.2%
  • Element/IonOil, Grease
  • FormLiquid
  • Volume (mL)10
  • Packaging20/PK
  • Storage SymbolStore between 15 and 30 °C
  • UN Number1090
  • CAS Number67-64-1, 57-11-4, 544-76-3
  • Shelf Life (months)24
  • DescriptionOil and Grease Snip™ Certified Reference Material, 0.2%, 10 mL; 20/PK
Documents & Certificates
Reviews
Q&A