NSI Lab Solutions PEO-020 PCB in Water Wastewater Proficiency Test; 2/PK

Representative image only.

NSI Lab Solutions PEO-020 PCB in Water Wastewater Proficiency Test; 2/PK

NSI Lab Solutions | Item: 6120243

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Key Features

  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • Each test includes all analytes required by the TNI Non-Potable Water (NPW) fields of testing
  • Supplied in duplicate for convenience and value
  • Each ampule produces at least 1 L of sample (except for VOCs)
  • ISO 17043 accredited
  • Two 1.5-mL concentrate set in acetone for use with Methods 608/8080/8081

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental chemistry wastewater proficiency tests are designed for identification of target chemicals in wastewater.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

For determination of the following elements:
Arochlor 1016: 2.0 to 10 µg/L
Arochlor 1221: 2.0 to 10 µg/L
Arochlor 1232: 2.0 to 10 µg/L
Arochlor 1242: 2.0 to 10 µg/L
Arochlor 1248: 2.0 to 10 µg/L
Arochlor 1254: 2.0 to 10 µg/L
Arochlor 1260: 2.0 to 10 µg/L
Arochlor 1262: 2.0 to 10 µg/L
Arochlor 1268: 2.0 to 10 µg/L

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • MatrixAcetone
  • Element/IonArochlor 1016, Arochlor 1221, Arochlor 1232, Arochlor 1242, Arochlor 1248, Arochlor 1254, Arochlor 1260, Arochlor 1268
  • FormLiquid
  • Volume (mL)1.5
  • Packaging2/PK
  • Storage SymbolRefrigerate between 2 and 8°C
  • UN Number1090
  • Shelf Life (months)24
  • DescriptionPCB in Water Wastewater Proficiency Test; 2/PK
Reviews
Q&A