NSI Lab Solutions PE-115 Diesel in Water Proficiency Test

Representative image only.

NSI Lab Solutions PE-115 Diesel in Water Proficiency Test

NSI Lab Solutions | Item: 6120305

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Key Features

  • Four studies per year without order deadlines
  • Convenient online reporting with PT Datalink
  • ISO 17043 accredited
  • A blind sample for dilution in water
  • Analyze for Diesel by Modified 8015 and NWTPH-Dx Methods in the range of 800 to 6000 µg/L
  • Single ampule

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental chemistry soil and hazardous waste proficiency tests are designed for identification of target chemicals in the soil and underground storage tanks (UST).

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • Element/IonDiesel
  • FormLiquid
  • Storage SymbolRefrigerate between 2 and 8 °C
  • UN Number1230
  • Shelf Life (months)24
  • DescriptionDiesel in Water Proficiency Test
Reviews
Q&A