NSI Lab Solutions MIC-UNV-20 Universal Water Microbe Mixed Culture Standard; 20/PK

Representative image only.

NSI Lab Solutions MIC-UNV-20 Universal Water Microbe Mixed Culture Standard; 20/PK

NSI Lab Solutions | Item: 0500340

Multiple organism CRM covers six methods in a single pellet!

Call For Price

Call For Price
Product Description

Key Features

  • Hydration fluid dissolves culture pellet in less than a minute
  • Compatible with MTF, IDEXX, and plate count methods—CoA includes expected value for multiple methods
  • Less than 10% RSD lot precision
  • No more than two passages from primary strain culture
  • ISO 17034 accredited
  • No royalty fee
  • Pack of 20

More About this Item

These microbiological reference materials are designed for quality control of water samples. Each dehydrated pellet contains multiple microorganisms that can be tested at once. The culture pellet lyophilized in Microgel-Flash™ matrix dissolves almost instantly with minimum agitation and no incubation. Values listed below are approximate activity levels; see the Certificate of Analysis (CoA) for actual values. Store in the freezer.

Microbiology Tests at Approximate Activity Levels:
Total Coliform: 2400 CFU/100 mL
E. coli: 1000 CFU/100 mL
Fecal Coliform: 500 CFU/100 mL
P. aeruginosa: 1000 CFU/100 mL
Enterococci: 1000 CFU/100 mL
HPC: 50 CFU/mL

Download Certificate of Analysis at nsilabsolutions.com/coa-downloads/.

Specifications
  • OrganismMultiple
  • Packaging20/PK
  • Product TypeEnvironmental Mixed Cultures
  • Shipping requirementFrozen ice pack
  • Storage requirementStore between –20 and –10°C
  • DescriptionUniversal Water Microbe Mixed Culture Standard; 20/PK
Reviews
Q&A