NSI Lab Solutions MIC-014 Quantitative Legionella Wastewater Proficiency Test

Representative image only.

NSI Lab Solutions MIC-014 Quantitative Legionella Wastewater Proficiency Test

NSI Lab Solutions | Item: 0500444

Ensure state and accreditation regulation compliance with proficiency testing

Call For Price

Call For Price
Product Description

Includes

Two bottles of sterile hydration fluid.

Key Features

  • Monthly testing without order deadlines
  • Convenient online reporting with PT Datalink
  • Supplied in duplicate for convenience and value
  • ISO 17043 accredited
  • Designed for use with Legiolert® or BCYE plate count methods
  • Sample supplied as a dehydrated pellet in the range of 20 to 2400 CFU as L. pneumophila

More About this Item

Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.

These environmental microbiology wastewater proficiency tests are designed for quantitative analysis of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.

If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.

Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.

Specifications
  • OrganismLegionella
  • Organism typeBacteria
  • Packaging2/PK
  • Product TypeWastewater Proficiency Test
  • Shelf Life Months (months)12
  • Shipping requirementFrozen ice packs
  • Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
  • DescriptionQuantitative Legionella Wastewater Proficiency Test
Reviews
Q&A