Designed for use with all Most Probable Numbers (MPN) and Membrane Filtration (MF) test procedures
Compatible with IDEXX Quanti-Tray® MPN methods for wastewater
Sample is supplied as a dehydrated pellet in the range of 20 to 2400 CFU/MPN per 100 mL (the NELAC analyte concentration range)
More About this Item
Use the NSI proficiency testing (PT) studies to help improve quality and comply with regulations. There are no limits on the number of analysts or number of methods that you may report. Receive your final reports within 21 days of the study close date.
These environmental microbiology wastewater proficiency tests are designed for quantitative analysis of target microorganisms. Each vacuum-sealed glass vial contains a culture pellet lyophilized in Microgel-Flash™ matrix. Simply hydrate with supplied hydration fluid. The culture pellet dissolves almost instantly with minimum agitation and no incubation.
If you have an immediate need for proficiency test results, PT Express™, our on-demand proficiency testing is available year-round. Call us at 1-800-234-7387 to place your order.
Lead time specified is for studies that have already opened. The lead time for studies that have not yet opened will be applicable once the study opens. To view study numbers and corresponding study dates, please see the 2024 Proficiency Testing Study Schedule linked under the Resources section below.
Specifications
OrganismColiforms, E.coli
Organism typeBacteria
Packaging2/PK
Product TypeWastewater Proficiency Test
Shelf Life Months (months)12
Shipping requirementFrozen ice packs
Storage requirementSample: store between –20 and –10°CHydration fluid: store at room temperature
DescriptionQuantitative Coliforms/E.coli Wastewater Proficiency Test